CO1H324A5F51 CombiPhos Catalysts Inc

5-(Imidazol-1-yl)pyridine-3-boronic acid

  • Cas Number : 2225180-68-5
  • Molecular Weight : 188.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart