CO1H324A5F52 CombiPhos Catalysts Inc

5-(2-Methylimidazol-1-yl)pyridine-3-boronic acid

  • Cas Number : 2225154-47-0
  • Molecular Weight : 203.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart