CO1H324A5F54 CombiPhos Catalysts Inc

5-(1H-Pyrrol-1-yl)pyridine-3-boronic acid

  • Cas Number : 2225178-11-8
  • Molecular Weight : 187.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart