CO1H324A5F5B CombiPhos Catalysts Inc

5-(3-Methylpiperidin-1-yl)pyridine-3-boronic acid

  • Cas Number : 2225153-52-4
  • Molecular Weight : 220.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart