CO1H324A5F67 CombiPhos Catalysts Inc

(2-(sec-butyl)pyridin-4-yl)boronic acid

  • Cas Number : 2225153-07-9
  • Molecular Weight : 179.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart