CO1H324A5F71 CombiPhos Catalysts Inc

5-Chloro-6-(iso-propyl)pyridine-3-boronic acid

  • Cas Number : 2225176-53-2
  • Molecular Weight : 199.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart