CO1H324A5F72 CombiPhos Catalysts Inc

4-Ethylpyridine-3-boronic acid

  • Cas Number : 1001907-67-0
  • Molecular Weight : 150.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart