CO1H324A5F73 CombiPhos Catalysts Inc

4-(n-Propyl)pyridine-3-boronic acid

  • Cas Number : 553652-71-4
  • Molecular Weight : 165

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart