CO1H324A5F7B CombiPhos Catalysts Inc

(2,5-dimethylpyridin-4-yl)boronic acid

  • Cas Number : 1788062-09-8
  • Molecular Weight : 150.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart