CO1H324A5F88 CombiPhos Catalysts Inc

5-Methoxypyridine-2-boronic acid

  • Cas Number : 163105-89-3
  • Molecular Weight : 152.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart