CO1H324A5F90 CombiPhos Catalysts Inc

2-(tert-Butyl)pyridine-3-boronic acid

  • Cas Number : 2225173-31-7
  • Molecular Weight : 179.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart