CO1H324A5F92 CombiPhos Catalysts Inc

2,3'-Bipyridine-3-boronic acid

  • Cas Number : 2225181-44-0
  • Molecular Weight : 200

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart