CO1H324A5F98 CombiPhos Catalysts Inc

2-(5-Pyrimidyl)pyridine-3-boronic acid

  • Cas Number : 2225154-92-5
  • Molecular Weight : 200.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart