CO1H324A5F9E CombiPhos Catalysts Inc

2-(4-Fluorophenyl)pyridine-3-boronic acid

  • Cas Number : 2225173-24-8
  • Molecular Weight : 217.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart