CO1H324A5FA4 CombiPhos Catalysts Inc

2-(4-Methoxyphenyl)pyridine-3-boronic acid

  • Cas Number : 2225177-98-8
  • Molecular Weight : 229.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart