CO1H324A5FA7 CombiPhos Catalysts Inc

2-(2-Methyl-4-fluorophenyl)pyridine-3-boronic acid

  • Cas Number : 2225174-40-1
  • Molecular Weight : 231.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart