CO1H324A5FAA CombiPhos Catalysts Inc

2-(4-Chlorophenyl)pyridine-3-boronic acid

  • Cas Number : 2225154-60-7
  • Molecular Weight : 233.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart