CO1H324A5FAB CombiPhos Catalysts Inc

2-(Thiazol-4-yl)pyridine-3-boronic acid

  • Cas Number : 2225173-30-6
  • Molecular Weight : 206.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart