CO1H324A5FB5 CombiPhos Catalysts Inc

(5-isobutylpyridin-3-yl)boronic acid

  • Cas Number : 1952356-61-4
  • Molecular Weight : 179.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart