CO1H324A5FB9 CombiPhos Catalysts Inc

[5-(AMINOMETHYL)PYRIDIN-3-YL]BORONIC ACID

  • Cas Number : 2225177-50-2
  • Molecular Weight : 151.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart