CO1H324A5FBA CombiPhos Catalysts Inc

(5-(piperidin-4-yl)pyridin-3-yl)boronic acid

  • Cas Number : 2225178-34-5
  • Molecular Weight : 206.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart