CO1H324A5FBD CombiPhos Catalysts Inc

4-(TRIFLUOROMETHYL)PYRIDINE-3-BORONIC ACID

  • Cas Number : 1072952-32-9
  • Molecular Weight : 190.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart