CO1H324A5FBE CombiPhos Catalysts Inc

5-(PROP-1-EN-2-YL)PYRIDINE-3-BORONIC ACID

  • Cas Number : 2225180-75-4
  • Molecular Weight : 162.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart