CO1H324A5FC6 CombiPhos Catalysts Inc

5-BROMO-6-(TRIFLUOROMETHYL)PYRIDIN-3-YLBORONIC ACID

  • Cas Number : 2225155-69-9
  • Molecular Weight : 269.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart