CO1H324A5FD5 CombiPhos Catalysts Inc

6-Cyclohexyl-5-trifluoromethylpyridine-3-boronic acid

  • Cas Number : 2225177-59-1
  • Molecular Weight : 273.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart