CO1H324A5FDA CombiPhos Catalysts Inc

6-BROMO-5-(METHOXYCARBONYL)PYRIDINE-3-BORONIC ACID

  • Cas Number : 2225154-74-3
  • Molecular Weight : 259.85

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart