CO1H324A5FDD CombiPhos Catalysts Inc

(2-CYANOPYRIDIN-4-YL)BORONIC ACID

  • Cas Number : 903513-60-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart