CO1H324A5FE0 CombiPhos Catalysts Inc

6-(2-TETRAHYDROFURANYL)PYRIDINE-3-BORONIC ACID

  • Cas Number : 2225153-42-2
  • Molecular Weight : 193.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart