CO1H324A5FE5 CombiPhos Catalysts Inc

(2-(piperidin-3-yl)pyrimidin-5-yl)boronic acid

  • Cas Number : 2225152-04-3
  • Molecular Weight : 207.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart