CO1H324A5FEE CombiPhos Catalysts Inc

6-Bromo-5-(cyclobutyl)pyridine-3-boronic acid

  • Cas Number : 2225181-47-3
  • Molecular Weight : 255.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart