CO1H324A5FEF CombiPhos Catalysts Inc

6-Bromo-5-(cyclopentyl)pyridine-3-boronic acid

  • Cas Number : 2225178-84-5
  • Molecular Weight : 269.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart