CO1H324A5FF0 CombiPhos Catalysts Inc

6-Bromo-5-(cyclohexyl)pyridine-3-boronic acid

  • Cas Number : 2225180-77-6
  • Molecular Weight : 283.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart