CO1H324A5FF6 CombiPhos Catalysts Inc

6-Chloro-5-ethylpyridine-3-boronic acid

  • Cas Number : 2225174-83-2
  • Molecular Weight : 185.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart