CO1H324A5FFA CombiPhos Catalysts Inc

6-Chloro-5-(cyclobutyl)pyridine-3-boronic acid

  • Cas Number : 2225181-48-4
  • Molecular Weight : 211.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart