CO1H324A5FFC CombiPhos Catalysts Inc

6-Chloro-5-(cyclohexyl)pyridine-3-boronic acid

  • Cas Number : 2225178-32-3
  • Molecular Weight : 239.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart