CO1H324A600A CombiPhos Catalysts Inc

6-Chloro-5-(4-trifluoromethylphenyl)pyridine-3-boronic acid

  • Cas Number : 2225179-27-9
  • Molecular Weight : 301.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart