CO1H324A600B CombiPhos Catalysts Inc

6-Chloro-5-(3-trifluoromethylphenyl)pyridine-3-boronic acid

  • Cas Number : 2225155-16-6
  • Molecular Weight : 301.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart