CO1H324A6018 CombiPhos Catalysts Inc

6-Chloro-5-(pyridin-3-yl)pyridine-3-boronic acid

  • Cas Number : 2225174-10-5
  • Molecular Weight : 234.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart