CO1H324A6022 CombiPhos Catalysts Inc

6-Chloro-5-(imidazol-1-yl)pyridine-3-boronic acid

  • Cas Number : 2225181-50-8
  • Molecular Weight : 223.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart