CO1H324A602D CombiPhos Catalysts Inc

5-Bromo-6-cyclobutoxypyridine-3-boronic acid

  • Cas Number : 2225177-70-6
  • Molecular Weight : 271.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart