CO1H324A6030 CombiPhos Catalysts Inc

5-Bromo-6-(cyclohexyloxy)pyridine-3-boronic acid

  • Cas Number : 2225175-13-1
  • Molecular Weight : 299.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart