CO1H324A6032 CombiPhos Catalysts Inc

5-Chloro-6-dimethylaminopyridine-3-boronic acid

  • Cas Number : 1704121-36-7
  • Molecular Weight : 200.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart