CO1H324A6042 CombiPhos Catalysts Inc

5-Chloro-6-(4-methylpiperidin-1-yl)pyridine-3-boronic acid

  • Cas Number : 2225177-79-5
  • Molecular Weight : 254.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart