CO1H324A6062 CombiPhos Catalysts Inc

5-Chloro-6-(4-methoxyphenyl)pyridine-3-boronic acid

  • Cas Number : 2225156-00-1
  • Molecular Weight : 263.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart