CO1H324A6069 CombiPhos Catalysts Inc

5-Chloro-6-(pyridin-3-yl)pyridine-3-boronic acid

  • Cas Number : 2225175-36-8
  • Molecular Weight : 234.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart