CO1H324A607D CombiPhos Catalysts Inc

(4-bromo-3-cyclopropylphenyl)boronic acid

  • Cas Number : 2225174-68-3
  • Molecular Weight : 240.89

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart