CO1H324A607E CombiPhos Catalysts Inc

(3-cyclopropyl-4-fluorophenyl)boronic acid

  • Cas Number : 2225178-16-3
  • Molecular Weight : 179.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart