CO1H324A6084 CombiPhos Catalysts Inc

(3-chloro-5-cyclopropylphenyl)boronic acid

  • Cas Number : 2225170-80-7
  • Molecular Weight : 196.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart