CO1H324A6085 CombiPhos Catalysts Inc

(3-bromo-5-cyclopropylphenyl)boronic acid

  • Cas Number : 2225155-28-0
  • Molecular Weight : 240.89

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart