CO1H324A6089 CombiPhos Catalysts Inc

(3-cyclopropyl-5-methoxyphenyl)boronic acid

  • Cas Number : 1787258-63-2
  • Molecular Weight : 192.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart